Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005672899.3 | XP_005672956.2 | 1024 | 91 | ||
XM_021078127.1 | XP_020933786.1 | 1059 | 91 | |||
XM_021078128.1 | XP_020933787.1 | 1055 | 91 | |||
XM_021078129.1 | XP_020933788.1 | 1053 | 91 | |||
XM_021078131.1 | XP_020933790.1 | 1068 | 91 | |||
GenBank mRNA | AK344514.1 | - | 8 - 9 | 922 | 91 | |
AY706384.1 | - | 7 - 8 | 987 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons |