Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206441.1 | NP_001193370.1 | 8 - 9 | 1611 | 77 | |
XM_021098372.1 | XP_020954031.1 | 1750 | 77 | |||
GenBank mRNA | AK396046.1 | - | 6 - 7 | 955 | 77 | |
DQ227991.1 | - | 7 - 8 | 1222 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |