Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128466.1 | NP_001121938.1 | 4 - 5 | 397 | 112 | |
XM_005666030.3 | XP_005666087.1 | 587 | 112 | |||
XM_021097971.1 | XP_020953630.1 | 771 | 112 | |||
GenBank mRNA | AK350063.1 | - | 5 - 6 | 558 | 112 | |
EU589332.1 | - | 4 - 5 | 397 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |