Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128468.1 | NP_001121940.1 | 3 - 4 | 342 | 66 | |
XM_021088474.1 | XP_020944133.1 | 479 | 66 | |||
GenBank mRNA | AK392462.1 | - | 3 - 4 | 548 | 66 | |
EU616815.1 | - | 3 - 4 | 342 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |