Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164007.1 | NP_001157479.1 | 7 - 8 | 1125 | 63 | |
GenBank mRNA | AK233241.1 | - | 7 - 8 | 1125 | 63 | |
AK392032.1 | - | 7 - 8 | 1110 | 63 | ||
M86719.1 | - | 7 - 8 | 883 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Inventoried Probe spans exons |