Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003482313.4 | XP_003482361.2 | 891 | 66 | ||
GenBank mRNA | Y17718.1 | - | 192 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa |