Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244121.1 | NP_001231050.1 | 3 | 1252 | 108 | |
GenBank mRNA | AK236774.1 | - | 3 | 1254 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon |