Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163404.1 | NP_001156876.1 | 5 - 6 | 711 | 75 | |
XM_021066215.1 | XP_020921874.1 | 1150 | 75 | |||
GenBank mRNA | AK236306.1 | - | 6 - 7 | 888 | 75 | |
AK391494.1 | - | 6 - 7 | 828 | 75 | ||
AK399443.1 | - | 6 - 7 | 834 | 75 | ||
AY010065.2 | - | 5 - 6 | 647 | 75 | ||
GQ166956.1 | - | 5 - 6 | 711 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |