Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC23A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003123979.5 | XP_003124027.1 | 1104 | 59 | ||
XM_013995181.2 | XP_013850635.1 | 1150 | 59 | |||
XM_013995183.2 | XP_013850637.1 | 1796 | 59 | |||
XM_021084869.1 | XP_020940528.1 | 858 | 59 | |||
XM_021084870.1 | XP_020940529.1 | 1150 | 59 | |||
GenBank mRNA | AK350105.1 | - | 7 | 1907 | 59 | |
AY353718.1 | - | 4 | 348 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |