Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY935636.1 | - | 1 - 2 | 71 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |