Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004036.2 | NP_001004036.1 | 2 | 151 | 83 | |
GenBank mRNA | V01303.1 | - | 152 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |