Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012299.2 | NP_001012299.1 | 2 | 1851 | 82 | |
GenBank mRNA | AY870646.1 | - | 2 | 1852 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |