Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCTEX1D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032356.1 | NP_001027527.1 | 3 | 746 | 138 | |
GenBank mRNA | AJ973122.1 | - | 3 | 746 | 138 | |
AK347503.1 | - | 3 | 758 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |