Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037149.1 | NP_001032226.1 | 2 | 248 | 97 | |
GenBank mRNA | AY731093.1 | - | 2 | 337 | 97 | |
AY953426.1 | - | 2 | 248 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |