Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038636.1 | NP_001033725.1 | 9 | 954 | 77 | |
XM_021082074.1 | XP_020937733.1 | 976 | 77 | |||
GenBank mRNA | DQ322459.1 | - | 9 | 954 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |