Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213761.1 | NP_998926.1 | 2 | 2352 | 83 | |
GenBank mRNA | AY392087.1 | - | 2 | 2352 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon |