Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF209702.1 | - | 4 - 5 | 450 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |