Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213864.1 | NP_999029.1 | 4 | 254 | 62 | |
GenBank mRNA | S46866.1 | - | 4 | 254 | 62 | |
X67109.1 | - | 254 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons |