Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213964.1 | NP_999129.1 | 67 | 9449 | 60 | |
XM_013978501.2 | XP_013833955.2 | 9415 | 60 | |||
XM_021100445.1 | XP_020956104.1 | 9298 | 60 | |||
XM_021100446.1 | XP_020956105.1 | 10295 | 60 | |||
XM_021100447.1 | XP_020956106.1 | 9550 | 60 | |||
GenBank mRNA | AB016793.1 | - | 67 | 9449 | 60 | |
AK394899.1 | - | 4 | 522 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |