Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214080.1 | NP_999245.1 | 2 | 250 | 113 | |
XM_021093514.1 | XP_020949173.1 | 3182 | 113 | |||
XM_021093517.1 | XP_020949176.1 | 2862 | 113 | |||
XM_021093518.1 | XP_020949177.1 | 2145 | 113 | |||
XM_021093519.1 | XP_020949178.1 | 1840 | 113 | |||
GenBank mRNA | AY329372.1 | - | 2 | 250 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |