Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL18R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214098.1 | NP_999263.1 | 10 | 1551 | 109 | |
XM_021085848.1 | XP_020941507.1 | 1920 | 109 | |||
XM_021085849.1 | XP_020941508.1 | 1840 | 109 | |||
XM_021085850.1 | XP_020941509.1 | 2112 | 109 | |||
XM_021085851.1 | XP_020941510.1 | 1631 | 109 | |||
GenBank mRNA | AB079258.1 | - | 10 | 1551 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |