Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214217.1 | NP_999382.1 | 3 | 1346 | 64 | |
XM_013980517.2 | XP_013835971.2 | 2072 | 64 | |||
GenBank mRNA | S78208.1 | - | 3 | 1346 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |