Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214391.2 | NP_999556.1 | 5 | 651 | 118 | |
XM_021088724.1 | XP_020944383.1 | 738 | 118 | |||
GenBank mRNA | AF372453.1 | - | 5 | 652 | 118 | |
AK346858.1 | - | 5 | 688 | 118 | ||
AK391241.1 | - | 5 | 694 | 118 | ||
KF595285.1 | - | 5 | 651 | 118 | ||
U08992.1 | - | 2 | 339 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons |