Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPNMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098584.1 | NP_001092054.1 | 11 | 1549 | 103 | |
XM_021101918.1 | XP_020957577.1 | 1861 | 103 | |||
XM_021101919.1 | XP_020957578.1 | 1569 | 103 | |||
GenBank mRNA | AB271936.1 | - | 11 | 1549 | 103 | |
AK230642.1 | - | 11 | 1623 | 103 | ||
AK230716.1 | - | 7 | 894 | 103 | ||
AK389619.1 | - | 11 | 1659 | 103 | ||
AK391303.1 | - | 11 | 1623 | 103 | ||
AK391325.1 | - | 12 | 1817 | 103 | ||
AK391342.1 | - | 6 | 664 | 103 | ||
AK391369.1 | - | 11 | 1616 | 103 | ||
AK391428.1 | - | 6 | 680 | 103 | ||
AK393455.1 | - | 2 | 383 | 103 | ||
AK399427.1 | - | 11 | 1623 | 103 | ||
AK399533.1 | - | 11 | 1515 | 103 | ||
AK399537.1 | - | 11 | 1623 | 103 | ||
AK399571.1 | - | 11 | 1659 | 103 | ||
AK400904.1 | - | 11 | 1616 | 103 |
Gene Symbol: LOC106508657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK391210.1 | - | 11 | 1622 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Amplicon spans exons and probe does not span exons |