Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105293.1 | NP_001098763.1 | 10 | 1045 | 109 | |
GenBank mRNA | EU046996.1 | - | 10 | 1045 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |