Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123175.1 | NP_001116647.1 | 6 | 541 | 69 | |
GenBank mRNA | AK396301.1 | - | 6 | 753 | 69 | |
DQ917634.1 | - | 6 | 541 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons |