Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126101.2 | NP_001119573.1 | 4 | 355 | 72 | |
GenBank mRNA | EU597631.1 | - | 4 | 355 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon |