Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044537.1 | NP_001038002.1 | 3 - 4 | 386 | 109 | |
GenBank mRNA | DQ325522.1 | - | 3 - 4 | 386 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Sus scrofa Probe spans exons |