Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044614.2 | NP_001038079.1 | 6 - 7 | 1248 | 93 | |
GenBank mRNA | AK349227.1 | - | 1 - 2 | 854 | 93 | |
AK389176.1 | - | 6 - 7 | 1262 | 93 | ||
AK395712.1 | - | 6 - 7 | 1289 | 93 | ||
AK395910.1 | - | 7 - 8 | 1343 | 93 | ||
AK396093.1 | - | 5 - 6 | 696 | 93 | ||
AK396682.1 | - | 3 - 4 | 2109 | 93 | ||
AK399333.1 | - | 2 - 3 | 429 | 93 | ||
AK400522.1 | - | 6 - 7 | 1260 | 93 | ||
AK400942.1 | - | 6 - 7 | 985 | 93 | ||
DQ365929.1 | - | 6 - 7 | 1249 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |