Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFIP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077214.1 | NP_001070682.1 | 4 - 5 | 572 | 81 | |
XM_021072205.1 | XP_020927864.1 | 637 | 81 | |||
XM_021072206.1 | XP_020927865.1 | 705 | 81 | |||
GenBank mRNA | DQ923315.1 | - | 4 - 5 | 572 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |