Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077215.1 | NP_001070683.1 | 12 - 13 | 1445 | 76 | |
XM_005660329.3 | XP_005660386.1 | 1713 | 76 | |||
XM_013993751.2 | XP_013849205.1 | 1749 | 76 | |||
XM_013993752.2 | XP_013849206.1 | 1684 | 76 | |||
XM_013993753.2 | XP_013849207.1 | 1677 | 76 | |||
XM_021084313.1 | XP_020939972.1 | 1668 | 76 | |||
GenBank mRNA | AK236015.1 | - | 13 - 14 | 1611 | 76 | |
AK236448.1 | - | 13 - 14 | 1653 | 76 | ||
AK343731.1 | - | 12 - 13 | 1606 | 76 | ||
DQ972969.1 | - | 12 - 13 | 1445 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |