Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077217.1 | NP_001070685.1 | 5 - 6 | 676 | 74 | |
GenBank mRNA | AK347966.1 | - | 5 - 6 | 814 | 74 | |
AK394817.1 | - | 5 - 6 | 815 | 74 | ||
AK397146.1 | - | 5 - 6 | 814 | 74 | ||
DQ972964.1 | - | 5 - 6 | 676 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |