Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077226.1 | NP_001070694.1 | 6 - 7 | 655 | 101 | |
XM_005664070.3 | XP_005664127.1 | 860 | 101 | |||
XM_013997827.2 | XP_013853281.1 | 1819 | 101 | |||
GenBank mRNA | AK348596.1 | - | 8 - 9 | 817 | 101 | |
AK396389.1 | - | 8 - 9 | 814 | 101 | ||
DQ972959.1 | - | 6 - 7 | 655 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |