Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPZA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097455.1 | NP_001090924.1 | 2 - 3 | 101 | 85 | |
GenBank mRNA | AK235584.1 | - | 2 - 3 | 201 | 85 | |
AK392663.1 | - | 2 - 3 | 216 | 85 | ||
AK399579.1 | - | 2 - 3 | 125 | 85 | ||
AK400274.1 | - | 2 - 3 | 315 | 85 | ||
AM410995.1 | - | 2 - 3 | 101 | 85 | ||
AY610172.1 | - | 2 - 3 | 195 | 85 | ||
EF202987.1 | - | 2 - 3 | 201 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |