Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097476.2 | NP_001090945.1 | 1 - 2 | 75 | 63 | |
GenBank mRNA | AK240209.1 | - | 1 - 2 | 80 | 63 | |
AK352187.1 | - | 1 - 2 | 62 | 63 | ||
AK391865.1 | - | 1 - 2 | 76 | 63 | ||
DQ629163.1 | - | 1 - 2 | 76 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |