Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097493.1 | NP_001090962.1 | 1 - 2 | 88 | 97 | |
XM_005660185.3 | XP_005660242.1 | 1736 | 97 | |||
GenBank mRNA | AK348760.1 | - | 1 - 2 | 209 | 97 | |
DQ439546.1 | - | 1 - 2 | 88 | 97 | ||
DQ439547.1 | - | 1 - 2 | 88 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |