Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097501.2 | NP_001090970.1 | 4 - 5 | 637 | 99 | |
XM_005659974.3 | XP_005660031.1 | 769 | 99 | |||
GenBank mRNA | AK232068.1 | - | 5 - 6 | 729 | 99 | |
AK350128.1 | - | 5 - 6 | 697 | 99 | ||
AK352387.1 | - | 5 - 6 | 702 | 99 | ||
EF053388.1 | - | 4 - 5 | 640 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |