Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097518.1 | NP_001090987.1 | 5 - 6 | 858 | 103 | |
XM_005654507.3 | XP_005654564.1 | 901 | 103 | |||
GenBank mRNA | AK232338.1 | - | 6 - 7 | 1573 | 103 | |
EF423616.1 | - | 5 - 6 | 858 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Sus scrofa Probe spans exons |