Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYD88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099923.1 | NP_001093393.1 | 2 - 3 | 466 | 119 | |
GenBank mRNA | AB292176.1 | - | 2 - 3 | 503 | 119 | |
EF198416.1 | - | 2 - 3 | 466 | 119 | ||
EU056736.1 | - | 2 - 3 | 515 | 119 | ||
EU077229.1 | - | 2 - 3 | 459 | 119 | ||
EU155133.1 | - | 2 - 3 | 289 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |