Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100195.1 | NP_001093665.1 | 9 - 10 | 1031 | 150 | |
GenBank mRNA | AK231721.1 | - | 9 - 10 | 1039 | 150 | |
AK391718.1 | - | 9 - 10 | 1031 | 150 | ||
DQ351285.1 | - | 9 - 10 | 1031 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |