Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101033.1 | NP_001094503.1 | 2 - 3 | 169 | 79 | |
XM_005672829.3 | XP_005672886.1 | 1668 | 79 | |||
XM_021077174.1 | XP_020932833.1 | 1668 | 79 | |||
GenBank mRNA | AK237864.1 | - | 2 - 3 | 175 | 79 | |
AK350647.1 | - | 2 - 3 | 186 | 79 | ||
DQ859019.1 | - | 2 - 3 | 169 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |