Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105306.1 | NP_001098776.1 | 5 - 6 | 1013 | 77 | |
GenBank mRNA | AK234056.1 | - | 5 - 6 | 1341 | 77 | |
AM397623.1 | - | 5 - 6 | 1013 | 77 | ||
DQ859925.1 | - | 5 - 6 | 1013 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |