Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109946.1 | NP_001103416.1 | 3 - 4 | 1281 | 62 | |
XM_005660427.3 | XP_005660484.1 | 1827 | 62 | |||
GenBank mRNA | AK235298.1 | - | 1 - 2 | 106 | 62 | |
EF154831.1 | - | 3 - 4 | 1281 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Inventoried Probe spans exons |