Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTCFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110174.1 | NP_001103644.1 | 2 - 3 | 751 | 66 | |
XM_013985484.2 | XP_013840938.1 | 766 | 66 | |||
XM_013985485.2 | XP_013840939.1 | 766 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |