Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110430.1 | NP_001103900.1 | 3 - 4 | 331 | 107 | |
XM_005673029.3 | XP_005673086.1 | 485 | 107 | |||
GenBank mRNA | AK235656.1 | - | 3 - 4 | 315 | 107 | |
AK239052.1 | - | 3 - 4 | 330 | 107 | ||
AK396372.1 | - | 3 - 4 | 345 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |