Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112684.1 | NP_001106154.1 | 3 - 4 | 380 | 87 | |
GenBank mRNA | AK344373.1 | - | 3 - 4 | 736 | 87 | |
AK391360.1 | - | 3 - 4 | 437 | 87 | ||
AK399607.1 | - | 3 - 4 | 692 | 87 | ||
EU194551.1 | - | 3 - 4 | 561 | 87 | ||
EU220427.1 | - | 3 - 4 | 380 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |