Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112686.1 | NP_001106156.1 | 4 - 5 | 623 | 144 | |
XM_021088448.1 | XP_020944107.1 | 951 | 144 | |||
XM_021088449.1 | XP_020944108.1 | 641 | 144 | |||
GenBank mRNA | AB304401.1 | - | 4 - 5 | 623 | 144 | |
AK235142.1 | - | 3 - 4 | 759 | 144 | ||
AK390167.1 | - | 4 - 5 | 863 | 144 | ||
AK390242.1 | - | 3 - 4 | 744 | 144 | ||
AK390347.1 | - | 4 - 5 | 858 | 144 | ||
AK390590.1 | - | 3 - 4 | 768 | 144 | ||
AY610416.1 | - | 4 - 5 | 551 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |