Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113287.1 | NP_001106758.1 | 6 - 7 | 811 | 94 | |
GenBank mRNA | AK232212.1 | - | 6 - 7 | 822 | 94 | |
AK239566.1 | - | 6 - 7 | 792 | 94 | ||
U07180.1 | - | 6 - 7 | 811 | 94 |
Gene Symbol: LDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013988347.2 | XP_013843801.1 | 881 | 94 | ||
XM_021092252.1 | XP_020947911.1 | 860 | 94 | |||
GenBank mRNA | AK389821.1 | - | 6 - 7 | 819 | 94 | |
AK391952.1 | - | 6 - 7 | 817 | 94 | ||
AK398230.1 | - | 6 - 7 | 817 | 94 | ||
AK400969.1 | - | 6 - 7 | 822 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |