Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113438.1 | NP_001106909.1 | 7 - 8 | 798 | 122 | |
XM_013980180.2 | XP_013835634.1 | 894 | 122 | |||
XM_013980181.2 | XP_013835635.1 | 833 | 122 | |||
XM_013980182.2 | XP_013835636.1 | 710 | 122 | |||
XM_013980183.2 | XP_013835637.1 | 690 | 122 | |||
XM_013980184.2 | XP_013835638.1 | 618 | 122 | |||
GenBank mRNA | AK390787.1 | - | 6 - 7 | 1211 | 122 | |
EU024116.1 | - | 7 - 8 | 798 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |