Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNISR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113439.1 | NP_001106910.1 | 8 - 9 | 1221 | 130 | |
XM_021079463.1 | XP_020935122.1 | 1973 | 130 | |||
XM_021079851.1 | XP_020935510.1 | 1271 | 130 | |||
XM_021080246.1 | XP_020935905.1 | 1152 | 130 | |||
XM_021080749.1 | XP_020936408.1 | 1152 | 130 | |||
GenBank mRNA | EU308569.1 | - | 8 - 9 | 1221 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |